2-Pyrrolidone 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyrrolidin-2-one

13211-0025 13211-2500 13211-5000
TS13211-0025EA 132 USD
TS13211-0025 TS13211-2500 TS13211-5000
2-Pyrrolidone 99%
2-Pyrrolidone
Formula: C₄H₇NO
Boiling Pt: 250 °C (760 mmHg)
Melting Pt: 25.5 °C
Density: 1.103 g/cm³
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00005270
CAS Number: 616-45-5
EINECS: 210-483-1
Merck Index: 12,08200

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid or low melting mass (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4860 to 1.4880 (20°C, 589 nm)
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR