Zinc chloride ∼6,5% (∼ 0,4Mol/L) in ethyl ether

Supplier: TCI America

Synonyms: Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride

Z0038-100ML
TCZ0038-100MLEA 107.12 USD
TCZ0038-100ML
Zinc chloride ∼6,5% (∼ 0,4Mol/L) in ethyl ether
Zinc chloride
Formula: ZnCl₂
MDL Number: MFCD00011295
CAS Number: 7646-85-7
UN: 2924

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR