Xanthene 98,0

Supplier: TCI America

Synonyms: 9H-Xanthene, Dibenzopyran

X0003-25G
TCX0003-025GEA 195.31 USD
TCX0003-025G
Xanthene 98,0
Xanthene

CAS Number: 92-83-1 MDL Number: MFCD00005055 Molecular Formula: C13H10O Molecular Weight: 182.22 Purity/Analysis Method: 98.0% (GC) Form: Crystal Color: White Boiling point (°C): 315 Melting point (°C): 100


Other Product Names / Synonyms: Dibenzopyran

Formula: C₁₃H₁₀O
MW: 182.22 g/mol
Boiling Pt: 315 °C
Melting Pt: 100 °C
MDL Number: MFCD00005055
CAS Number: 92-83-1
EINECS: 202-194-4

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR