Tetrabutylammonium hydroxide 37% in methanol

Supplier: TCI America

T1948-25ML T1948-500ML
TCT1948-25MLEA 37.65 USD
TCT1948-25ML TCT1948-500ML
Tetrabutylammonium hydroxide 37% in methanol
Tetrabutylammonium hydroxide

CAS Number: 2052-49-5
MDL Number: MFCD00009425
Molecular Formula: C16H37NO
Molecular Weight: 259.48
Form: Clear Liquid
Color: Colorless
Flash Point (°C): 11

Formula: C₁₆H₃₇NO
Flash Pt: 11 °C
MDL Number: MFCD00009425
CAS Number: 2052-49-5
UN: 3263
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e75978a90d30b3709eaf134d63773e77bb784e9&vid=72616a069e7d9e80dbe462d1cbcf5430b4412a43&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR