1,4,7-Trimethyl-1,4,7-triazacyclononane ≥98.0% stabilised

Supplier: TCI America

Synonyms: 1,4,7-Trimethyl-1,4,7-triazonane

T1879-1G T1879-5G
TCT1879-1GEA 289.8 USD
TCT1879-1G TCT1879-5G
1,4,7-Trimethyl-1,4,7-triazacyclononane ≥98.0% stabilised
1,4,7-Trimethyl-1,4,7-triazacyclononane

(stabilized with NaHCO3) CAS Number: 96556-05-7 Molecular Formula: C9H21N3 Molecular Weight: 171.29 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Color: Colorless Flash Point (°C): 68 Specific Gravity (20/20): 0.90 Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Ligands, Materials Chemistry, Precise Polymerization Reagents, Macrocycles for Host-Guest Chemistry Other Product Names / Synonyms: Octahydro-1,4,7-trimethyl-1H-1,4,7-triazonine

Formula: C₉H₂₁N₃
Density: 0.90 (20 °C)
Flash Pt: 68 °C
MDL Number: MFCD00012359
CAS Number: 96556-05-7
UN: 3267
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3e0c32d86e515828e13f479cfc6b9c9ba6d8bb5&vid=c3e0c32d86e515828e13f479cfc6b9c9ba6d8bb5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR