Tetrakis(triphenylphosphine)palladium(0) ≥97.0%

Supplier: TCI America

Synonyms: Tetrakis(triphenylphosphine)palladium, Pd(PPh3)4

T1350-1G T1350-25G T1350-5G
TCT1350-001GEA 38 USD
TCT1350-001G TCT1350-25G TCT1350-005G
Tetrakis(triphenylphosphine)palladium(0) ≥97.0%
Tetrakis(triphenylphosphine)palladium (0)

CAS Number: 14221-01-3 MDL Number: MFCD00010012 Molecular Formula: C72H60P4Pd Molecular Weight: 1155.59 Purity/Analysis Method: 97.0% (T) Form: Crystal Color: Pale Yellow Melting point (°C): 105 Storage Temperature: -20°C

Formula: C₇₂H₆₀P₄Pd
Melting Pt: 105 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00010012
CAS Number: 14221-01-3
EINECS: 238-086-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=42507881e5f2d0a3f882862dad5ac1a78f3e6dd7&vid=42507881e5f2d0a3f882862dad5ac1a78f3e6dd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR