Trichloromethanesulfonyl chloride ≥95.0%

Supplier: TCI America

T0611-25G T0611-5G
TCT0611-025GEA 301 USD
TCT0611-025G TCT0611-005G
Trichloromethanesulfonyl chloride ≥95.0%
Trichloromethanesulfonyl chloride

CAS Number: 2547-61-7 MDL Number: MFCD00007452 Molecular Formula: CCl4O2S Molecular Weight: 217.87 Purity/Analysis Method: 95.0% (T) Form: Crystal Color: White Melting point (°C): 141

Formula: CCl₄O₂S
MW: 217.87 g/mol
Melting Pt: 141 °C
MDL Number: MFCD00007452
CAS Number: 2547-61-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4358125bc14528b34abaaabcd24a39e02253612c&vid=4358125bc14528b34abaaabcd24a39e02253612c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR