L-Selenomethionine ≥98.0%

Supplier: TCI America

Synonyms: L-SelenoMethionine, (S)-2-Amino-4-(methylseleno)butyric Acid, H-Se-Met-OH

S0442-1G S0442-5G
TCS0442-001GEA 185.38 USD
TCS0442-001G TCS0442-5G
L-Selenomethionine ≥98.0%
L-Selenomethionine

CAS Number: 3211-76-5 MDL Number: MFCD00037210 Molecular Formula: C5H11NO2Se Molecular Weight: 196.11 Purity/Analysis Method: 98.0% (HPLC,T) Form: Crystal Color: White Melting point (°C): 267 Specific rotation [a]20/D: 18 deg (C=0.5, 2mol/L HCl) Storage Temperature: 0°C


Product Categories: Bioscience, Antioxidants, Biologically-modified Amino Acids Other Product Names / Synonyms: (S)-2-Amino-4-(methylseleno)butyric Acid

Formula: C₅H₁₁NO₂Se
Melting Pt: 267 °C
MDL Number: MFCD00037210
CAS Number: 3211-76-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eaf4a41ad5b3aee547baf4b51859343375397f2&vid=6eaf4a41ad5b3aee547baf4b51859343375397f2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR