Phenyl trifluoromethanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Phenyl Trifluoromethanesulphonate, Trifluoromethanesulphonic acid Phenyl Ester, Phenyl Trifluoromethanesulfonate, Phenyl Triflate, Trifluoromethanesulfonic acid Phenyl Ester

P1829-5G P1829-1G
TCP1829-5GEA 156 USD
TCP1829-5G TCP1829-1G
Phenyl trifluoromethanesulfonate ≥98.0%
Phenyl trifluoromethanesulfonate

CAS Number: 17763-67-6 MDL Number: MFCD00192399 Molecular Formula: C7H5F3O3S Molecular Weight: 226.17 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Color: Colorless Boiling point (°C): 62 Flash Point (°C): 71 Specific Gravity (20/20): 1.41


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives Other Product Names / Synonyms: Trifluoromethanesulfonic Acid Phenyl Ester, Phenyl Triflate

Formula: C₇H₅F₃O₃S
MW: 226.17 g/mol
Boiling Pt: 62 °C
Density: 1.41 (20 °C)
Flash Pt: 71 °C
MDL Number: MFCD00192399
CAS Number: 17763-67-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82b7f070a0c7558b4c65c5891574163e3283f100&vid=82b7f070a0c7558b4c65c5891574163e3283f100&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR