Murexide

Supplier: TCI America

Synonyms: Ammonium purpurate, Murexide triturate, Ammonium 5-(2,4,6-trioxoperhydropyrimidin-5-ylideneamino)barbiturate, Purpuric acid ammonium salt

M0474-1G M0474-5G M0474-25G
TCM0474-001GEA 25 USD
TCM0474-001G TCM0474-5G TCM0474-025G
Murexide
Murexide

CAS Number: 3051-09-0 MDL Number: MFCD00012777 Molecular Formula: C8H8N6O6 Molecular Weight: 284.19 Form: Crystal Color: Deep Red


Other Product Names / Synonyms: Ammonium Purpurate

Formula: C₈H₈N₆O₆
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00012777
CAS Number: 3051-09-0
EINECS: 221-266-6
Merck Index: 13,06330
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b93691b6275560c0c5bf6d44836c4a8871128ea6&vid=b93691b6275560c0c5bf6d44836c4a8871128ea6&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR