3-Methylpyrocatechol ≥99.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3-Methylcatechol, 3-Methylbenzene-1,2-diol

M0184-25G M0184-5G
TCM0184-025GEA 273 USD
TCM0184-025G TCM0184-5G
3-Methylpyrocatechol ≥99.0% (by GC)
3-Methylpyrocatechol

CAS Number: 488-17-5 MDL Number: MFCD00016435 Molecular Formula: C7H8O2 Molecular Weight: 124.14 Purity/Analysis Method: 99.0% (GC) Form: Crystal Boiling point (°C): 125 Melting point (°C): 66


Other Product Names / Synonyms: 1,2-Dihydroxy-3-methylbenzene, 2,3-Dihydroxytoluene, 3-Methylpyrocatechol

Formula: C₇H₈O₂
Boiling Pt: 125 °C
Melting Pt: 66 °C
MDL Number: MFCD00016435
CAS Number: 488-17-5
EINECS: 207-672-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3dec0bc24158ac9e86649f4da816f83dd0ad4b1d&vid=ee05f9bf14d87efe9cacca17fffdc20e74b00ef7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 99.0 %
Melting point 64.0 to 68.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR