Lead(II) acetate ≥98.0% (by titrimetric analysis) for perovskite precursor

Supplier: TCI America
Danger

Synonyms: Acetic acid Lead(II) salt

L0315-1G L0315-25G L0315-5G
TCL0315-1GEA 23 USD
TCL0315-1G TCL0315-25G TCL0315-5G
Lead(II) acetate ≥98.0% (by titrimetric analysis) for perovskite precursor
Lead (II) acetate
Formula: (H₃CCOO)₂Pb
Melting Pt: 280 °C
MDL Number: MFCD00012452
CAS Number: 301-04-2
EINECS: 206-104-4
UN: 1616
ADR: 6.1,III
Merck Index: 14,05397

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Chelometric Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR