1-Hexanesulfonic acid sodium salt ≥98.0%

Supplier: TCI America

Synonyms: Sodium hexane-1-sulfonate, Sodium hexane-1-sulphonate, Sodium hexanesulfonate, Sodium hexanesulphonate, Sodium 1-hexanesulfonate, Sodium 1-hexanesulphonate

I0344-25G I0344-5G I0344-100G
TCI0344-025GEA 171.75 USD
TCI0344-025G TCI0344-005G TCI0344-100G
1-Hexanesulfonic acid sodium salt ≥98.0%
1-Hexanesulphonic acid sodium salt

[Reagent for Ion-Pair Chromatography] CAS Number: 2832-45-3 MDL Number: MFCD00007542 Molecular Formula: C6H14O3S Molecular Weight: 188.22 Purity/Analysis Method: 98.0% (T) Form: Crystal


Product Categories: Analytical Science, HPLC Ion-Pair Reagents for Basic Samples Other Product Names / Synonyms: 1-Hexanesulfonic Acid Sodium Salt, IPC-ALKS-6

Formula: C₆H₁₃O₃SNa
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007542
CAS Number: 2832-45-3
EINECS: 220-601-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbd900e15e6b0bcc5b73728e8a7e3b0bebee0aa8&vid=d56895c8dd24c4a50cde58a68b6c9bdaf5edf5e6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR