Imidazole ≥99.0%

Supplier: TCI America
Danger

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

I0288-25G I0288-100G
TCI0288-25GEA 177 USD
TCI0288-25G TCI0288-100G
Imidazole ≥99.0%
Imidazole

[for Buffer] CAS Number: 288-32-4 MDL Number: MFCD00005183 Molecular Formula: C3H4N2 Molecular Weight: 68.08 Purity/Analysis Method: 99.0% (GC,T) Form: Crystal Boiling point (°C): 256 Melting point (°C): 90 Flash Point (°C): 145

Formula: C₃H₄N₂
Boiling Pt: 256 °C
Melting Pt: 90 °C
Density: 1.18 g/cm³ (20 °C)
Flash Pt: 145 °C
Storage Temperature: Ambient
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 2923
ADR: 8,III
Merck Index: 13,04935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea5798efeb4994381f38b277e69ed3e78f73b8a5&vid=d3a9cb8feb6ffd7cc8b0d8bceb34c9fbc1960d10&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR