N,N-Diethylhydroxylamine ≥98.0%

Supplier: TCI America

D1339-500ML D1339-25ML D1339-100ML
TCD1339-500MLEA 102 USD
TCD1339-500ML TCD1339-25ML TCD1339-100ML
N,N-Diethylhydroxylamine ≥98.0%
N,N-Diethylhydroxylamine

CAS Number: 3710-84-7 MDL Number: MFCD00002126 Molecular Formula: C4H11NO Molecular Weight: 89.14 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 122 Flash Point (°C): 51 Specific Gravity (20/20): 0.87

Formula: C₄H₁₁NO
Boiling Pt: 122 °C
Density: 0.87 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00002126
CAS Number: 3710-84-7
EINECS: 223-055-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5983be6139da5517027b116de8c1253f058dc6b9&vid=5983be6139da5517027b116de8c1253f058dc6b9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR