Sodium 1-decanesulfonate ≥98.0%

Supplier: TCI America

Synonyms: Sodium decane-1-sulphonate, Decane-1-sulphonic acid sodium salt, 1-Decanesulphonic acid, sodium salt, 1-Decanesulfonic acid, sodium salt

D1181-25G D1181-5G
TCD1181-025GEA 84.42 USD
TCD1181-025G TCD1181-5G
Sodium 1-decanesulfonate ≥98.0%
Sodium 1-decanesulphonate

CAS Number: 13419-61-9 MDL Number: MFCD00007526 Molecular Formula: C10H22O3S Molecular Weight: 244.32 Purity/Analysis Method: 98.0% (T) Form: Crystal


Other Product Names / Synonyms: 1-Decanesulfonic Acid Sodium Salt

Formula: C₁₀H₂₁NaO₃S
Melting Pt: > 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00007526
CAS Number: 13419-61-9
EINECS: 236-525-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6225ac831d49953c0f9e1877c73cefe0c3616bd&vid=e6225ac831d49953c0f9e1877c73cefe0c3616bd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR