2-Cyclopentylethanol ≥98.0%

Supplier: TCI America

C0817-5ML C0817-25ML
TCC0817-005MLEA 192.5 USD
TCC0817-005ML TCC0817-25ML
2-Cyclopentylethanol ≥98.0%
2-Cyclopentylethanol

CAS Number: 766-00-7 MDL Number: MFCD00019301 Molecular Formula: C7H14O Molecular Weight: 114.19 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 182 Flash Point (°C): 70 Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: 2-Cyclopentylethanol, 2-Hydroxyethylcyclopentane

Formula: C₇H₁₄O
Boiling Pt: 182 °C
Density: 0.92 (20 °C)
Flash Pt: 70 °C
MDL Number: MFCD00019301
CAS Number: 766-00-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d5fe8661f0f574d1d26ccca338784c6a85a88c2&vid=8d5fe8661f0f574d1d26ccca338784c6a85a88c2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR