Chloroform ≥99.0% stabilised

Supplier: TCI America
Danger

Synonyms: Methane, trichloro-, Trichloroform, Methylidyne trichloride, Trichloromethane

Stabilization: Stabilised with Stabilised

C0175-100ML
TCC0175-100MLEA 62.99 USD
TCC0175-100ML
Chloroform ≥99.0% stabilised
Chloroform

(stabilized with Ethanol) [for Spectrophotometry] CAS Number: 67-66-3 MDL Number: MFCD00000826 Molecular Formula: CHCl3 Molecular Weight: 119.37 Purity/Analysis Method: 99.0% (GC) Form: Clear Liquid Boiling point (°C): 60 Melting point (°C): -64 Specific Gravity (20/20): 1.49

Formula: CHCl₃
Boiling Pt: 60 °C
Melting Pt: –64 °C
Density: 1.49 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
EINECS: 200-663-8
UN: 1888
ADR: 6.1,III
REACH: 05-2114614906-45
Merck Index: 13,02160
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bc707049706522f7759b9757791c070f9087adc9&vid=2543bf493a3633fe2f94bd8a0a53ba97eaca0f71&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR