4-Bromoanisole ≥97.0%

Supplier: TCI America

B0547-25G B0547-500G B0547-100G
TCB0547-025GEA 26.84 USD
TCB0547-025G TCB0547-500G TCB0547-100G
4-Bromoanisole ≥97.0%
4-Bromoanisole

CAS Number: 104-92-7 MDL Number: MFCD00000097 Molecular Formula: C7H7BrO Molecular Weight: 187.04 Purity/Analysis Method: 97.0% (GC) Form: Clear Liquid Boiling point (°C): 215 Flash Point (°C): 94 Freezing point (°C): 12 Specific Gravity (20/20): 1.50


Other Product Names / Synonyms: 1-Bromo-4-methoxybenzene

Formula: C₇H₇BrO
MW: 187.04 g/mol
Boiling Pt: 223
Melting Pt: 10
Density: 1.50 (20 °C)
Flash Pt: 94
Storage Temperature: Ambient
MDL Number: MFCD00000097
CAS Number: 104-92-7
EINECS: 203-252-1
Merck Index: 13,01412
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=859125b799f9f3e651e9034406cca25a7e66b0e3&vid=d8869bf90f325b7c5cd0164e9799fb17f86d2306&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR