Benzalkonium chloride 50% in water

Supplier: TCI America
Danger

Synonyms: Quaternary ammonium compounds, benzyl-C8-18-alkyldimethyl, chlorides, Alkylbenzyldimethylammonium chloride

B0414-500G B0414-25G
TCB0414-500GEA 117.87 USD
TCB0414-500G TCB0414-25G
Benzalkonium chloride 50% in water
Benzalkonium chloride

CAS Number: 63449-41-2
MDL Number: MFCD00050250
Form: Clear Liquid
Color: Colorless


Other Product Names / Synonyms: Alkyldimethylbenzylammonium Chloride, Benzyldimethylalkylammonium Chloride

Formula: C₆H₅CH₂N(CH₃)₂RCl (R=C₈H₁₇ to C₁₈H₃₇)
Storage Temperature: Ambient
MDL Number: MFCD00145757
CAS Number: 63449-41-2
EINECS: 264-151-6
UN: 2923
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=27f05fb34530410e60259100ac06710026903603&vid=846e9d9b588e1d5fafc5feacfe6443dcd6154a9c&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR