Claritas PPT® Grade ICP-MS Multi-Element Solution 1, SPEX CertiPrep
Supplier: SPEX CERTIPREP LLC
CLMS-1
SXCLMS-1EA 445.44 USD
SXCLMS-1
Claritas PPT® Grade ICP-MS Multi-Element Solution 1, SPEX CertiPrep
Standards Cationic Standards Multi-Element Standards
Claritas PPT® multi-element standards for ICP-MS.
SPEX CertiPrep Claritas PPT® standards are a class of Inorganic Certified Reference Standards designed specifically for today's new generation of trace ICP and ICP-MS instrumentation. Based on extensive development, our chemists have formulated this line of high-purity standards for user convenience and stability.
The Claritas PPT® selection of standards includes a complete series of multi-element solutions, many designed for use with US EPA Methods. These solutions are made with the highest purity materials available and are tested on our state-of-the-art ICP-MS.
Multi-Solution Solution Standards: Designed to contain virtually every element in the mass spectrum for concentration verification checks.
Certifications: SPEX CertiPrep Certified Reference Materials (CRMs) are manufactured under a quality system complying with the requirements of ISO 9001, ISO/IEC 17025 and ISO 17034.