Sodium thiosulfate 0.0394 N, Carbon Black reagent for ASTM D1510

Supplier: SPEX CERTIPREP LLC

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

182002
SX182002EA 125.15 USD
SX182002
Sodium thiosulfate 0.0394 N, Carbon Black reagent for ASTM D1510
Sodium thiosulphate

Sodium thiosulfate is a Carbon Black Reagent designed for ASTM D1510.


Our sodium thiosulfate solutions are prepared for ACS Grade, micro-crystalline Na₂S₂O₃. In order to maximize shelf life, our matrix is prepared using double-deionized, ASTM Type I Water.


All solutions are prepared gravimetrically using high accuracy analytical balances to ensure precise target concentrations. Each batch is thoroughly homogenized using a high speed industrial mixer to ensure reliable results from the first bottle to the last.

Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
MDL Number: MFCD00003499
CAS Number: 7772-98-7
Merck Index: 12,08844

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR