BL21 Competent Cells, Agilent Technologies
Supplier: Agilent Technologies
200131 200132 200133 230130 230132 230134 230240 230245 230255 230280
97066-330PK 349.99 USD
97066-330 99900-860 ST200131 ST200132 ST200133 ST230130 ST230132 ST230134 ST230240 ST230245 ST230255 ST230275 76193-370
BL21 Competent Cells, Agilent Technologies
Competent Cells
BL21 competent cells are an all-purpose strain for high-level protein expression and easy induction.
  • High-level protein expression and easy induction
  • Can be used with non-T7 RNA polymerase protein expression systems
  • Allows the tightest control of protein expression for extremely toxic proteins
  • Suitable for expressing proteins from all species, including mammals
  • Ideal for difficult protein expression, especially when codon bias is a problem
  • Increases transformation efficiency with Hte phenotype
  • Derived from Agilent’s high performance BL21-Gold competent
More Product Information