Ammonium thiocyanate 0,5% w/v methanol_dilute_sulfuric acid

Supplier: Ricca Chemical

Synonyms: Thiocyanic acid ammonium salt, Ammonium rhodanide, Ammonium sulphocyanate, Ammonium sulfocyanate

RSOD5500-4A
RCSOD55-1EA 870 USD
RCSOD55-1
Ammonium thiocyanate 0,5% w/v methanol_dilute_sulfuric acid
Ammonium thiocyanate
Formula: NH₄SCN
MW: 76.12 g/mol
Melting Pt: 150
MDL Number: MFCD00011428
CAS Number: 1762-95-4
Merck Index: 13,00564

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR