VeriSpec™ Nitrite(N) - NO₂₋ f(N) Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Supplier: Ricca Chemical
RV010932-100N RV010932-250N RV010932-500N
RCRV010932100NEA 108 USD
RCRV010932100N RCRV010932250N RCRV010932500N
VeriSpec™ Nitrite(N) - NO₂₋ f(N) Standard for Ion Chromatography 1000 ppm in H₂O, RICCA Chemical Company
Standards Anionic Standards Nitrite Standards
Certified reference material (CRM) for ion chromatography (IC)
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility