VeriSpec™ Nexion AFT ICP-MS Detection Solution, 13 Components, RICCA Chemical Company
Supplier: Ricca Chemical
RV010889-100N
RCRV010889100NEA 429 USD
RCRV010889100N
VeriSpec™ Nexion AFT ICP-MS Detection Solution, 13 Components, RICCA Chemical Company
Standards Cationic Standards Multi-Element Standards
Certified reference material (CRM) for ICP-MS
50 ppb: Al, Ba, Ce, Co, In, Li, Mg, Mn, Ni, Pb, Tb, U, Zn
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility