VeriSpec™ Gallium Standard for ICP/MS 10 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010589-100N
RCRV010589100NEA 140 USD
RCRV010589100N
VeriSpec™ Gallium Standard for ICP/MS 10 ppm in 2% HNO₃
Standards Cationic Standards Gallium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility