VeriSpec™ Dysprosium Standard for ICP/MS 10 ppm in 2% HNO₃
Supplier: Ricca Chemical
RV010585-100N
RCRV010585100NEA 735 USD
RCRV010585100N
VeriSpec™ Dysprosium Standard for ICP/MS 10 ppm in 2% HNO₃
Standards Cationic Standards Dysprosium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility