VeriSpec® Bismuth Standard for ICP/MS 10 ppm in 2% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RV010570-100N
RCRV010570100NEA 130 USD
RCRV010570100N
VeriSpec® Bismuth Standard for ICP/MS 10 ppm in 2% HNO₃, Ricca Chemical Company
Standards Cationic Standards Bismuth Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility