VeriSpec™ Zirconium Standard for ICP/MS 100 ppm in 2% HNO₃/HF, RICCA Chemical Company
Supplier: Ricca Chemical
RV010569-100N
RCRV010569100NEA 159 USD
RCRV010569100N
VeriSpec™ Zirconium Standard for ICP/MS 100 ppm in 2% HNO₃/HF, RICCA Chemical Company
Standards Cationic Standards Zirconium Standards
Certified reference material (CRM) for ICP-MS
Certifications: Manufactured and Tested in an ISO 17025/ISO 17034 Accredited Facility