Zinc 0.1 mol/L

Supplier: Ricca Chemical

R9448500-500A
RCR9448500500AEA 237 USD
RCR9448500500A
Zinc 0.1 mol/L
Zinc
Formula: Zn
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6

Order Now

Specification Test Results

Appearance Colorless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR