Lead solution_stock 100 ppm Pb

Supplier: Ricca Chemical
Warning

R4296000-500A
RCR4296000500AEA 215 USD
RCR4296000500A
Lead solution_stock 100 ppm Pb
Lead
Formula: Pb
MW: 207.2 g/mol
Boiling Pt: 1740
Melting Pt: 327
Storage Temperature: Ambient
MDL Number: MFCD00134050
CAS Number: 7439-92-1
Merck Index: 12,05410

Order Now

Specification Test Results

Appearance Colorless liquid
Lead (Pb) 99.5-100.5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR