Hydrofluoric acid 2% v/v

Supplier: Ricca Chemical
Danger

R3819200-1A
RCR3819200-1AEA 290 USD
RCR3819200-1A
Hydrofluoric acid 2% v/v
Hydrofluoric acid
Formula: HF
MW: 20.01
Storage Temperature: Ambient
MDL Number: MFCD00011346
CAS Number: 7664-39-3
UN: 1790
ADR: 8 (6.1) II

Order Now

Specification Test Results

Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 1.93-2.07 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR