Ferric chloride 9% (w/v) in aqueous solution USP Test Solution (TS)

Supplier: Ricca Chemical
Danger

Synonyms: Iron(III) chloride solution

R3109000-500C R3109000-120C
RC3109-16EA 114 USD
RC3109-16 RCR3109000120C
Ferric chloride 9% (w/v) in aqueous solution USP Test Solution (TS)
Iron(III) chloride solutions
Formula: FeCl₃·6H₂O
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.1000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00149712
CAS Number: 10025-77-1
UN: 2582
ADR: 8,III

Order Now

Specification Test Results

Appearance Bright orange/brown liquid
Assay (vs. Sodium Thiosulfate/Starch) 8.9-9.1 % (w/v)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR