Ethylene glycol, Reagent Grade

Supplier: Ricca Chemical
Danger

Synonyms: Monoethylene glycol, Ethane-1,2-diol, 2-Hydroxyethanol, Ethylene alcohol, Ethanediol, Glycol alcohol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, 1,2-Dihydroxyethane, Glycol, Dihydroxyethane

R2944500-20F R2944500-4C R2944500-500C
RCR2944500-20FEA 728 USD
RCR2944500-20F RCR2944500-4C RCR2944500500C
Ethylene glycol, Reagent Grade
Ethylene glycol
Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: ∼ 197°C
Melting Pt: -13
Density: 1.1135
Flash Pt: 111°C
MDL Number: MFCD00002885
CAS Number: 107-21-1
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless viscous liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR