Nitric acid 5% v/v acid blank

Supplier: Ricca Chemical
Danger

Synonyms: Nitric acid solution

RPNACID5-500N RPNACID5-1N
RCPNACID5-500EA 139 USD
RCPNACID5-500 RCPNACID5-1L
Nitric acid 5% v/v acid blank
Nitric acid
Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.51
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Specification Test Results

Aluminum (Al) Actual Value Reported
Antimony (Sb) Actual Value Reported
Arsenic (As) Actual Value Reported
Barium (Ba) Actual Value Reported
Beryllium (Be) Actual Value Reported
Bismuth (Bi) Actual Value Reported
Boron (B) Actual Value Reported
Cadmium (Cd) Actual Value Reported
Calcium (Ca) Actual Value Reported
Cerium (Ce) Actual Value Reported


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR