Aluminum ICP Standard, 1 ml = 10 mg Al (10000 ppm Al)Al(NO₃)₃ in 5% HNO₃, Ricca Chemical Company
Supplier: Ricca Chemical
RPAL10KN-100N RPAL10KN-50N RPAL10KN-500N
RCPAL10KN-100EA 223 USD
RCPAL10KN-100 RCPAL10KN-50 RCPAL10KN-500
Aluminum ICP Standard, 1 ml = 10 mg Al (10000 ppm Al)Al(NO₃)₃ in 5% HNO₃, Ricca Chemical Company
Standards Cationic Standards Aluminum Standards