Nickel (5,0% Ni) nitric acid 5% Matrix_Modifier_Solution

Supplier: Ricca Chemical

RMNI50KN-50N
RCMMNI50KN-50EA 531 USD
RCMMNI50KN-50
Nickel (5,0% Ni) nitric acid 5% Matrix_Modifier_Solution
Nickel
Formula: Ni
MW: 58.69 g/mol
Boiling Pt: 3007
Melting Pt: 1453
MDL Number: MFCD00011137
CAS Number: 7440-02-0
UN: 3089
ADR: 4.1 III
Merck Index: 12,06582

Order Now

Specification Test Results

Appearance Clear, light green, odorless
Assay 50,000 ± 5000 ppm Ni


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR