Magnesium nitrate (2,0% Mg) nitric acid 5% Matrix_Modifier_Solution

Supplier: Ricca Chemical

RMMG20KN-50N
RCMMMG20KN-50EA 415 USD
RCMMMG20KN-50
Magnesium nitrate (2,0% Mg) nitric acid 5% Matrix_Modifier_Solution
Magnesium nitrate solutions
Formula: Mg(NO₃)₂
CAS Number: 10377-60-3
UN: 1474
ADR: 5.1 III

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Certified Concentration 20,000 ± 1000 ppm Mg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR