Water ACS, reagent analytical reagent ASTM Type I, ASTM Type II Packaged in glass containers

Supplier: Ricca Chemical

Synonyms: Dihydrogen oxide

R9151000-4C
RC91511CS 260 USD
RC91511
Water ACS, reagent analytical reagent ASTM Type I, ASTM Type II Packaged in glass containers
Water

Packaged in glass for applications requiring extremely low levels of organics. This product is manufactured by a continuous process that has been validated to produce high purity Water meeting, as a minimum, the specifications below at the time of packaging. Although this water is not distilled, it has a chemical purity equal to or better than double distilled water.
This product also meets ASTM Type III and Type IV specifications.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Colorless liquid
Chloride (Cl) max 1 ppb
Conductivity at 25°C max 0.056 µS/cm
Heavy Metals (as Pb) max 0.01 ppm
Nitrate (NO₃) max 0.4 ppm
Organic Carbon (TOC) max 50 ppb
Oxidizable Substances (Permanganate Retention) To Pass Test
Phosphate (PO₄) max 1.0 ppm
Silicate (as SiO₂) max 3 ppb
Sodium (Na) max 1 ppb
Sulfate (SO₄) max 1.0 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR