Sodium hypochlorite (6% Cl₂) in aqueous solution environmental

Supplier: Ricca Chemical

7495.7-1 7495.7-32
RC7495.7-1EA 328 USD
RC7495.7-1 RC7495.7-32
Sodium hypochlorite (6% Cl₂) in aqueous solution environmental
Sodium hypochlorite
Formula: ClNaO
MDL Number: MFCD00011120
CAS Number: 7681-52-9
UN: 1791
ADR: 8,II

Order Now

Specification Test Results

Assay 6 ± 0.06 % (w/w)
NIST SRM 136


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR