Silver nitrate 0.100 N (N/10) in aqueous solution analytical reagent

Supplier: Ricca Chemical
Warning

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

R7000000-4B R7000000-500B R7000000-10F R7000000-20F R7000000-1B R7000000-120B
RC70001EA 624 USD
RC70001 RC700016 RC700025 RC70005 RC700032 RC7000-4
Silver nitrate 0.100 N (N/10) in aqueous solution analytical reagent
Silver nitrate

Standardized at 25 °C against chemicals whose certification is traceable to NIST.

Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
EINECS: 231-853-9
UN: 3082
ADR: 9 III
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 999) 0.1000 ± 0.0001 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR