Potassium hydroxide 15% w/v aqueous solution

Supplier: Ricca Chemical

Synonyms: Potash caustic, Potassium hydrate, Caustic potash

R6135000-20F
RC61355EA 511 USD
RC61355
Potassium hydroxide 15% w/v aqueous solution
Potassium hydroxide
Formula: KOH
MW: 56.11 g/mol
Boiling Pt: 1320
Melting Pt: 360
MDL Number: MFCD00003553
CAS Number: 1310-58-3
UN: 1813
ADR: 8 II
Merck Index: 12,07806

Order Now

Specification Test Results

% (w/v) (traceable to NIST SRM 723) 15.0 ± 0.1 % (w/v)
Appearance Clear, colorless liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR