Phenolphthalein 0,5% (w/v) in 50% (v/v) alcohol indicator, neutralized

Supplier: Ricca Chemical
Danger

Synonyms: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one

R5600000-4A R5600000-500A R5600000-1A R5600000-120A
RC56001EA 188 USD
RC56001 RC560016 RC560032 RC56004
Phenolphthalein 0,5% (w/v) in 50% (v/v) alcohol indicator, neutralized
Phenolphthalein
Formula: C₂₀H₁₄O₄
MW: 318.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00005913
CAS Number: 77-09-8
EINECS: 201-004-7
UN: 1170
ADR: 3 III
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, very light pink, alcohol odor
Suitability for Use Colorless (pH 8.0) - Pink (pH 10.0)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR