Nitric acid 2.00 N analytical reagent

Supplier: Ricca Chemical
Danger

Synonyms: Nitric acid solution

R5432000-1A R5432000-500A R5432000-4A
RC5432-32EA 95 USD
RC5432-32 RC5432-16 RCR5432000-4A
Nitric acid 2.00 N analytical reagent
Nitric acid
Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.035 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 3264
ADR: 8 III
Merck Index: 13,06610

Order Now

Specification Test Results

Appearance Colorless to light yellow liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 1.999-2.001 N at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR