Hydrochloric acid 10% (v/v) in aqueous solution (1 + 9) analytical reagent

Supplier: Ricca Chemical
Warning

Synonyms: Hydrochloric acid solution, Muriatic acid

R3570000-20F R3570000-4A R3570000-500A R3570000-1A
RC3570-5EA 247 USD
RC3570-5 RC35701 RC357016 RC357032
Hydrochloric acid 10% (v/v) in aqueous solution (1 + 9) analytical reagent
Hydrochloric acid
Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear, colorless, odorless
Assay at 20 °C (traceable to NIST SRM 84) 10.00 ± 0.08 % (v/v)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR