Glycerine 50% v/v aqueous solution (1 + 1)

Supplier: Ricca Chemical
Warning

Synonyms: Glyceritol, Glysanin, 1,2,3-Propanetriol, Osmoglyn, Glycerin, Glycyl alcohol, Glyrol, Propane-1,2,3-triol, Propanetriol, Trihydroxypropane, Glycerol

3290-32
RC3290-32EA 93 USD
RC3290-32
Glycerine 50% v/v aqueous solution (1 + 1)
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.1420 g/cm³
Flash Pt: 177
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Specific Gravity 1.14 ± 0.005 at 20°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR