Ammonium iron (II) sulfate 0.00282 M (0.00282 N) 1 mL = 0.1 mg Cl2

Supplier: Ricca Chemical

R3144000-1C R3144000-4C R3144000-500C
RC314432EA 97 USD
RC314432 RC3144-1 RC3144-16
Ammonium iron (II) sulfate 0.00282 M (0.00282 N) 1 mL = 0.1 mg Cl2
Haematoxylin Harris

1 ml = 0.1 mg Cl₂

Formula: (NH₄)₂Fe(SO₄)₂·6H₂O
MW: 392.14 g/mol
Boiling Pt: 100 °C
Melting Pt: 0.0 °C
Density: 1.0010 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00150530
CAS Number: 7783-85-9

Order Now

Specification Test Results

Appearance Clear, light green, odorless
Assay at 20 °C (traceable to NIST SRM 136) 0.002820 ± 0.000005 N at 20.0 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR