Copper Standard, 1 ml = 10 mg Cu, 10,000 ppm Cu, Ricca Chemical Company
Supplier: Ricca Chemical
R2305000-500A R2305000-120A
RC230516EA 259 USD
RC230516 RC23054
Copper Standard, 1 ml = 10 mg Cu, 10,000 ppm Cu, Ricca Chemical Company
Standards Cationic Standards Copper Standards
Order Now
SPECIFICATIONS
Environmentally Preferable