Ceric ammonium sulfate 0.100 N (N/10) in 1 N sulfuric acid

Supplier: Ricca Chemical
Danger

Synonyms: DIAMMONIUM CERIUM(4+) TRIsulphATE, DIAMMONIUM CERIUM(4+) TRISulphATE, DIAMMONIUM CERIUM(4+) TRISULFATE, Ammonium cerium(4+) sulfate (2:1:3), Ammonium cerium(4+) sulphate (2:1:3), DIAMMONIUM CERIUM(4+) TRIsulfATE

1890-1 1890-32
RC1890-1EA 252 USD
RC1890-1 RC1890-32
Ceric ammonium sulfate 0.100 N (N/10) in 1 N sulfuric acid
Ceric ammonium sulfate
Formula: H₈CeN₂O₁₂S₃
MW: 464.381 Da
Boiling Pt: ~ 100 °C
Melting Pt: ~ 0 °C
Density: 1.0300 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00148851
CAS Number: 13840-04-5

Order Now

Specification Test Results

Assay 0.1 ± 0.0001 N at 20 °C
NIST SRM 83


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR